3,4-Dihydro-4-methyl-2H-pyrido[3,2-b][1,4]oxazine-7-carboxylic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Carboxy-3,4-dihydro-4-methyl-2H-pyrido[3,2-b][1,4]oxazine, 4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carboxylic acid

OR12220-250MG
APOSOR12220-250MGEA 202 EUR
APOSOR12220-250MG
3,4-Dihydro-4-methyl-2H-pyrido[3,2-b][1,4]oxazine-7-carboxylic acid 95+%
3,4-Dihydro-4-methyl-2H-pyrido[3,2-b][1,4]oxazine-7-carboxylic acid
Formel: C₉H₁₀N₂O₃
Molecular Weight: 194.187 g/mol
Schmelzpunkt: 231.5…280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09065017
CAS-Nummer: 915707-58-3
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR