1-(Thieno[3,2-d]pyrimidin-4-yl)piperidine-4-carboxylic acid 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Carboxy-1-(thieno[3,2-d]pyrimidin-4-yl)piperidine, 4-(4-Carboxypiperidin-1-yl)thieno[3,2-d]pyrimidine

OR12221-5G
APOSOR12221-5GEA 244 EUR
APOSOR12221-5G
1-(Thieno[3,2-d]pyrimidin-4-yl)piperidine-4-carboxylic acid 97%
1-(Thieno[3,2-d]pyrimidin-4-yl)piperidine-4-carboxylic acid
Formel: C₁₂H₁₃N₃O₂S
Molecular Weight: 263,316 g/mol
Schmelzpunkt: 247,5…249 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09065023
CAS-Nummer: 910037-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR