{4-[(Tetrahydro-2H-pyran-4-yl)oxy]phenyl}methylamine 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-[(Tetrahydro-2H-pyran-4-yl)oxy]benzylamine, 4-[4-(Aminomethyl)phenoxy]tetrahydro-2H-pyran

OR12223-250MG
APOSOR12223-250MGEA 48 EUR
APOSOR12223-250MG
{4-[(Tetrahydro-2H-pyran-4-yl)oxy]phenyl}methylamine 97%
{4-[(Tetrahydro-2H-pyran-4-yl)oxy]phenyl}methylamine
Formel: C₁₂H₁₇NO₂
Molecular Weight: 207.269 g/mol
Schmelzpunkt: 41.5…43.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09064967
CAS-Nummer: 864266-61-5
UN: 3259
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR