(2-Chlor-4-pyridinyl)methanol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Chloropyridin-4-yl)methanol, 2-Chloro-4-(hydroxymethyl)pyridine, 2-Chloropyridine-4-methanol

OR12246-5G
APOSOR12246-5GEA 46.4 EUR
APOSOR12246-5G
(2-Chlor-4-pyridinyl)methanol 97%
(2-Chlor-4-pyridinyl)methanol
Formel: C₆H₆ClNO
Molecular Weight: 143.57 g/mol
Schmelzpunkt: 59…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06858767
CAS-Nummer: 100704-10-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR