3-[3-(Dimethylamino)propoxy]benzyl alcohol 97%

Lieferant: Apollo Scientific

OR12248-1G
APOSOR12248-1GEA 0 EUR
APOSOR12248-1G
3-[3-(Dimethylamino)propoxy]benzyl alcohol 97%
3-[3-(Dimethylamino)propoxy]benzyl alcohol
Formel: C₁₂H₁₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09064989
CAS-Nummer: 912569-56-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR