(4-[3-(Dimethylamino)propoxy]phenyl)methanol

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-[3-(Dimethylamino)propoxy]benzyl alcohol

OR12249-1G
APOSOR12249-1GEA 78.3 EUR
APOSOR12249-1G
(4-[3-(Dimethylamino)propoxy]phenyl)methanol
(4-[3-(Dimethylamino)propoxy]phenyl)methanol
Formel: C₁₂H₁₉NO₂
Molecular Weight: 209,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06202771
CAS-Nummer: 426831-08-5
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR