5-Bromo-4-methyl-1H-imidazole

Lieferant: Apollo Scientific
Achtung

OR1225-1G
APOSOR1225-1GEA 46.4 EUR
APOSOR1225-1G
5-Bromo-4-methyl-1H-imidazole
5-Bromo-4-methyl-1H-imidazole
Formel: C₄H₅BrN₂
Molecular Weight: 161,0000 g/mol
Schmelzpunkt: 140…143 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06659898
CAS-Nummer: 15813-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR