5-Methylisoxazole-3-carbonitrile 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Cyano-5-methylisoxazole

OR12255-100MG
APOSOR12255-100MGEA 21.9 EUR
APOSOR12255-100MG
5-Methylisoxazole-3-carbonitrile 97%
5-Methylisoxazole-3-carbonitrile
Formel: C₅H₄N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09064940
CAS-Nummer: 57351-99-2
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR