1-Methyl-1H-pyrazole-3-carbonitrile 97%

Lieferant: Apollo Scientific
Achtung

OR12256-100MG
APOSOR12256-100MGEA 24.8 EUR
APOSOR12256-100MG
1-Methyl-1H-pyrazole-3-carbonitrile 97%
1-Methyl-1H-pyrazole-3-carbonitrile
Formel: C₅H₅N₃
Molecular Weight: 107.12 g/mol
Schmelzpunkt: 41.5…45 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00466343
CAS-Nummer: 79080-39-0
UN: 3439
ADR: 6.1 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR