3-(Pyrimidin-2-yl)benzylamine 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-[3-(Aminomethyl)phenyl]pyrimidine, 2-[3-(Aminomethyl)phenyl]-1,3-diazine

OR12261-1G
APOSOR12261-1GEA 277 EUR
APOSOR12261-1G
3-(Pyrimidin-2-yl)benzylamine 97%
3-(Pyrimidin-2-yl)benzylamine
Formel: C₁₁H₁₁N₃
Molecular Weight: 185,225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09702384
CAS-Nummer: 910036-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR