3-[3-(Dimethylamino)propoxy]benzyl chloride hydrochloride 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-[3-(Chloromethyl)phenoxy]-N,N-dimethylpropylamine hydrochloride

OR12271-250MG
APOSOR12271-250MGEA 186 EUR
APOSOR12271-250MG
3-[3-(Dimethylamino)propoxy]benzyl chloride hydrochloride 95%
3-[3-(Dimethylamino)propoxy]benzyl chloride hydrochloride
Formel: C₁₂H₁₈ClNO·ClH
Molecular Weight: 264.191 g/mol
Schmelzpunkt: 100.5…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09702394
CAS-Nummer: 926921-62-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR