2-Chloro-4-(fur-2-yl)pyrimidine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-4-(furan-2-yl)pyrimidine, 2-(2-Chloropyrimidin-4-yl)furan

OR12277-250MG
APOSOR12277-250MGEA 58 EUR
APOSOR12277-250MG
2-Chloro-4-(fur-2-yl)pyrimidine 97%
2-Chloro-4-(fur-2-yl)pyrimidine
Formel: C₈H₅ClN₂O
Molecular Weight: 180,591 g/mol
Schmelzpunkt: 81,5…82.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09065030
CAS-Nummer: 124959-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR