2,4-Dibrom-1-methylimidazol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,4-Dibrom-1-methyl-1H-imidazol
OR1228-100MG
APOSOR1228-100MGEA
88.6
EUR
APOSOR1228-100MG
2,4-Dibrom-1-methylimidazol 98%
2,4-Dibromo-1-methylimidazole
Formel:
C₄H₄Br₂N₂ Molecular Weight: 239,9 g/mol Schmelzpunkt: 74…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659906 CAS-Nummer: 53857-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...