5-Methyl-2-thiophencarbonitril 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Methylthiophene-2-carbonitrile, 2-Cyano-5-methylthiophene

OR12283-5G
APOSOR12283-5GEA 71.2 EUR
APOSOR12283-5G
5-Methyl-2-thiophencarbonitril 95%
5-Methyl-2-thiophencarbonitril
Formel: C₆H₅NS
Molecular Weight: 123.1756 g/mol
Siedepunkt: 73…75 °C/5mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD01434334
CAS-Nummer: 72835-25-7
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR