5-(Aminomethyl)benzo[b]furan 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-(Aminomethyl)-1-benzofuran, (Benzo[b]furan-5-yl)methylamine
OR12314-1G
OR12314-250MG
APOSOR12314-1GEA
346
EUR
APOSOR12314-1G
APOSOR12314-250MG
5-(Aminomethyl)benzo[b]furan 97%
5-(Aminomethyl)benzo[b]furan
Formel:
C₉H₉NO Schmelzpunkt: 32…34 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04974039 CAS-Nummer: 37798-08-6 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...