(4-Methyl-2-phenyl-1,3-thiazol-5-yl)methylamine 95%

Lieferant: Apollo Scientific
Gefahr

OR12318-1G
APOSOR12318-1GEA 226 EUR
APOSOR12318-1G
(4-Methyl-2-phenyl-1,3-thiazol-5-yl)methylamine 95%
(4-Methyl-2-phenyl-1,3-thiazol-5-yl)methylamine
Formel: C₁₁H₁₂N₂S
Lagertemperatur: Kühlschrank
MDL: MFCD05865105
CAS-Nummer: 165735-95-5
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR