tert-Butyl-4-(4-cyanophenyl)tetrahydro-1(2H)-pyrazinecarboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR12333-1G
OR12333-5G
APOSOR12333-1GEA
21.9
EUR
APOSOR12333-1G
APOSOR12333-5G
tert-Butyl-4-(4-cyanophenyl)tetrahydro-1(2H)-pyrazinecarboxylate 97%
tert-Butyl-4-(4-cyanophenyl)tetrahydro-1(2H)-pyrazinecarboxylate
Formel:
C₁₆H₂₁N₃O₂ Schmelzpunkt: 123…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03791198 CAS-Nummer: 186650-98-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...