Indoline-2-carboxylic acid, N-BOC protected 90%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(tert-Butoxycarbonyl)indoline-2-carboxylic acid, 1-(tert-Butoxycarbonyl)-2,3-dihydro-1H-indole-2-carboxylic acid, 1-(tert-Butoxycarbonyl)-2-carboxyindoline
OR12334-250MG
APOSOR12334-250MGEA
21.9
EUR
APOSOR12334-250MG
Indoline-2-carboxylic acid, N-BOC protected 90%
Indoline-2-carboxylic acid, N-BOC protected
Formel:
C₁₄H₁₇NO₄ Schmelzpunkt: 121…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02682411 CAS-Nummer: 137088-51-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...