tert-Butyl-4-(2-nitrophenoxy)tetrahydro-1(2H)-pyridinecarboxylate

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-(2-nitrophenoxy)piperidine-1-carboxylate, 4-(2-Nitrophenoxy)piperidine, N-BOC protected

OR12335-1G
APOSOR12335-1GEA 217 EUR
APOSOR12335-1G
tert-Butyl-4-(2-nitrophenoxy)tetrahydro-1(2H)-pyridinecarboxylate
tert-Butyl-4-(2-nitrophenoxy)tetrahydro-1(2H)-pyridinecarboxylate
Formel: C₁₆H₂₂N₂O₅
Molecular Weight: 322,36 g/mol
Schmelzpunkt: 88…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05865133
CAS-Nummer: 690632-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR