Quinoxaline-2-carbaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Formylquinoxaline

OR12342-1G
APOSOR12342-1GEA 37.7 EUR
APOSOR12342-1G
Quinoxaline-2-carbaldehyde 95%
Quinoxaline-2-carbaldehyde
Formel: C₉H₆N₂O
Molecular Weight: 158,16 g/mol
Schmelzpunkt: 107…111 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00272280
CAS-Nummer: 1593-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR