3,4-Dihydro-2H-1,4-benzoxazine-2-carbonitrile 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR12346-100MG
APOSOR12346-100MGEA
21.9
EUR
APOSOR12346-100MG
3,4-Dihydro-2H-1,4-benzoxazine-2-carbonitrile 97%
3,4-Dihydro-2H-1,4-benzoxazine-2-carbonitrile
Formel:
C₉H₈N₂O Schmelzpunkt: 64…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200862 CAS-Nummer: 86267-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...