1-Benzyl-5-methyl-1H-1,2,3-triazole-4-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Benzyl-4-carboxy-5-methyl-1H-1,2,3-triazole, [(4-Carboxy-5-methyl-1h-1,2,3-triazol-1-yl)methyl]benzene

OR12355-1G
APOSOR12355-1GEA 0 EUR
APOSOR12355-1G
1-Benzyl-5-methyl-1H-1,2,3-triazole-4-carboxylic acid 97%
1-Benzyl-5-methyl-1H-1,2,3-triazole-4-carboxylic acid
Formel: C₁₁H₁₁N₃O₂
Molecular Weight: 217,224 g/mol
Schmelzpunkt: 94…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06411557
CAS-Nummer: 54698-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR