Methyl-2-chloro-6-methylpyrimidine-4-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-chloro-6-methylpyrimidine-4-carboxylate

OR1236-5G
APOSOR1236-5GEA 183 EUR
APOSOR1236-5G
Methyl-2-chloro-6-methylpyrimidine-4-carboxylate
Methyl-2-chloro-6-methylpyrimidine-4-carboxylate
Formel: C₇H₇ClN₂O₂
Molecular Weight: 186,6 g/mol
Schmelzpunkt: 108…113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00117906
CAS-Nummer: 89793-11-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR