[2-(Thiomorpholin-4-yl)pyridin-4-yl]methanol
Lieferant: Apollo Scientific
Synonyme:
4-(Hydroxymethyl)-2-(thiomorpholin-4-yl)pyridine, 4-[4-(Hydroxymethyl)pyridin-2-yl]thiomorpholine
OR12361-1G
APOSOR12361-1GEA
198
EUR
APOSOR12361-1G
[2-(Thiomorpholin-4-yl)pyridin-4-yl]methanol
[2-(Thiomorpholin-4-yl)pyridin-4-yl]methanol
Formel:
C₁₀H₁₄N₂OS Molecular Weight: 210,296 g/mol Schmelzpunkt: 102…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09702362 CAS-Nummer: 898289-25-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...