4-(1-Methyl-1H-pyrazol-3-yl)benzonitril 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR12364-250MG
APOSOR12364-250MGEA
27.7
EUR
APOSOR12364-250MG
4-(1-Methyl-1H-pyrazol-3-yl)benzonitril 97%
4-(1-Methyl-1H-pyrazol-3-yl)benzonitril
Formel:
C₁₁H₉N₃ Schmelzpunkt: 146…147 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09702350 CAS-Nummer: 915707-41-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...