[(5-Methyl-2-thienyl)methyl]amine hydrochloride 97%

Lieferant: Apollo Scientific
Gefahr

OR12377-100MG
APOSOR12377-100MGEA 0 EUR
APOSOR12377-100MG
[(5-Methyl-2-thienyl)methyl]amine hydrochloride 97%
[(5-Methyl-2-thienyl)methyl]amine hydrochloride
Formel: C₆H₁₀ClNS
Schmelzpunkt: 191…193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09817548
CAS-Nummer: 171661-55-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR