6-Chloro-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(6-Chloro-3-oxo-2H-benzo[b][1,4]oxazin-4(3H)-yl)acetic acid

OR1238-1G
APOSOR1238-1GEA 368 EUR
APOSOR1238-1G
6-Chloro-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid
6-Chloro-3-oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid
Formel: C₁₀H₈ClNO₄
Molecular Weight: 241,6300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665821
CAS-Nummer: 26494-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR