3-[(6-Methylpyrazin-2-yl)oxy]benzoesäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR12386-250MG
APOSOR12386-250MGEA
198
EUR
APOSOR12386-250MG
3-[(6-Methylpyrazin-2-yl)oxy]benzoesäure 97%
3-[(6-Methylpyrazin-2-yl)oxy]benzoesäure
Formel:
C₁₂H₁₀N₂O₃ Schmelzpunkt: 130…134 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
906352-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...