4-(Pyrimidin-2-yl)benzenesulphonyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-[(4-Chlorosulphonyl)phenyl]pyrimidine, 2-[(4-Chlorosulphonyl)phenyl]-1,3-diazine

OR12391-250MG
APOSOR12391-250MGEA 1020 EUR
APOSOR12391-250MG
4-(Pyrimidin-2-yl)benzenesulphonyl chloride 97%
4-(Pyrimidin-2-yl)benzenesulphonyl chloride
Formel: C₁₀H₇ClN₂O₂S
Molecular Weight: 254,693 g/mol
Schmelzpunkt: 159,5…161.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09817488
CAS-Nummer: 912569-53-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR