[6-(Pyrrolidin-1-yl)pyridin-2-yl]methanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR12404-250MG
APOSOR12404-250MGEA
55.1
EUR
APOSOR12404-250MG
[6-(Pyrrolidin-1-yl)pyridin-2-yl]methanol 97%
[6-(Pyrrolidin-1-yl)pyridin-2-yl]methanol
Formel:
C₁₀H₁₄N₂O Schmelzpunkt: 44…46,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09702357 CAS-Nummer: 868755-48-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...