1-Methyl-1H-1,3-benzimidazole-5-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methyl-1H-benzo[d]imidazole-5-carboxylic acid

OR12418-5G
APOSOR12418-5GEA 154 EUR
APOSOR12418-5G
1-Methyl-1H-1,3-benzimidazole-5-carboxylic acid 97%
1-Methyl-1H-1,3-benzimidazole-5-carboxylic acid
Formel: C₉H₈N₂O₂
Molecular Weight: 176.18 g/mol
Schmelzpunkt: 293…295 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06809665
CAS-Nummer: 53484-17-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR