1-Methyl-1H-benzimidazole-5-carbonyl chloride hydrochloride

Lieferant: Apollo Scientific
Gefahr

OR12421-250MG
APOSOR12421-250MGEA 174 EUR
APOSOR12421-250MG
1-Methyl-1H-benzimidazole-5-carbonyl chloride hydrochloride
1-Methyl-1H-benzimidazole-5-carbonyl chloride hydrochloride
Formel: C₉H₇ClN₂O·ClH
Molecular Weight: 231,079 g/mol
Schmelzpunkt: 227,5…231.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09817460
CAS-Nummer: 906352-57-6
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR