3-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoyl chloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR12422-250MG
APOSOR12422-250MGEA
154
EUR
APOSOR12422-250MG
3-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoyl chloride 97%
3-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoyl chloride
Formel:
C₁₀H₇ClN₂O₂ Molecular Weight: 222,628 g/mol Schmelzpunkt: 82…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09817467 CAS-Nummer: 915707-46-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...