Methyl 4-hydroxyquinazoline-7-carboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydroxy-7-(methoxycarbonyl)quinazoline, 4-Hydroxy-7-(methoxycarbonyl)-1,3-benzodiazine
OR12426-1G
APOSOR12426-1GEA
72.5
EUR
APOSOR12426-1G
Methyl 4-hydroxyquinazoline-7-carboxylate 97%
Methyl 4-hydroxyquinazoline-7-carboxylate
Formel:
C₁₀H₈N₂O₃ Molecular Weight: 204.182 g/mol Schmelzpunkt: 252…254 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00681283 CAS-Nummer: 313535-84-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...