3-(1-Methyl-1H-pyrazol-3-yl)benzenesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-[3-(Chlorosulphonyl)phenyl]-1-methyl-1H-pyrazole

OR12428-1G
APOSOR12428-1GEA 0 EUR
APOSOR12428-1G
3-(1-Methyl-1H-pyrazol-3-yl)benzenesulphonyl chloride
3-(1-Methyl-1H-pyrazol-3-yl)benzenesulphonyl chloride
Formel: C₁₀H₉ClN₂O₂S
Molecular Weight: 256,709 g/mol
Schmelzpunkt: 76,5…78.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09817503
CAS-Nummer: 912569-59-6
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR