1-(Thieno[3,2-d]pyrimidin-4-yl)piperidine-4-carboxaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Formyl-1-(thieno[3,2-d]pyrimidin-4-yl)piperidine, 4-(4-Formylpiperidin-1-yl)thieno[3,2-d]pyrimidine, 1-(Thieno[3,2-d]pyrimidin-4-yl)piperidine-4-carbaldehyde

OR12436-250MG
APOSOR12436-250MGEA 63.8 EUR
APOSOR12436-250MG
1-(Thieno[3,2-d]pyrimidin-4-yl)piperidine-4-carboxaldehyde 97%
1-(Thieno[3,2-d]pyrimidin-4-yl)piperidine-4-carboxaldehyde
Formel: C₁₂H₁₃N₃OS
Molecular Weight: 247,316 g/mol
Schmelzpunkt: 94…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09817540
CAS-Nummer: 916766-91-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR