5-Amino-1-phenyl-1H-pyrazole-4-carboxamide

Lieferant: Apollo Scientific
Achtung

OR12451-250MG
APOSOR12451-250MGEA 21.9 EUR
APOSOR12451-250MG
5-Amino-1-phenyl-1H-pyrazole-4-carboxamide
5-Amino-1-phenyl-1H-pyrazole-4-carboxamide
Formel: C₁₀H₁₀N₄O
Molecular Weight: 202,22 g/mol
Schmelzpunkt: 167…170 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00020732
CAS-Nummer: 50427-77-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR