2-(3,4-Dichloroanilino)-1,3-thiazole-4-carboxylic acid

Lieferant: Apollo Scientific

OR12461-1G
APOSOR12461-1GEA 336 EUR
APOSOR12461-1G
2-(3,4-Dichloroanilino)-1,3-thiazole-4-carboxylic acid
2-(3,4-Dichloroanilino)-1,3-thiazole-4-carboxylicacid
Formel: C₁₀H₆Cl₂N₂O₂S
Molecular Weight: 289,14 g/mol
Schmelzpunkt: 289…291 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09817453
CAS-Nummer: 952182-44-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR