Ethyl-2-cyanobutanoate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-cyanobutanoate, Ethyl 2-cyanobutyrate

OR12466-25G
APOSOR12466-25GEA 0 EUR
APOSOR12466-25G
Ethyl-2-cyanobutanoate 98%
Ethyl-2-cyanobutanoate
Formel: C₇H₁₁NO₂
Molecular Weight: 141.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02258598
CAS-Nummer: 1619-58-5
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR