Methyl amino(3,4-dichlorophenyl)acetate hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: 3,4-Dichloro-DL-phenylglycine methyl ester hydrochloride

OR12471-1G OR12471-250MG
APOSOR12471-1GEA 452 EUR
APOSOR12471-1G APOSOR12471-250MG
Methyl amino(3,4-dichlorophenyl)acetate hydrochloride 95%
Methyl amino(3,4-dichlorophenyl)acetate hydrochloride
Formel: C₉H₁₀Cl₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04115512
CAS-Nummer: 1078611-21-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR