3-Phenyl-5,6-dihydroimidazo[2,1-b][1,3]thiazole-2-carbaldehyde

Lieferant: Apollo Scientific

OR12476-500MG
APOSOR12476-500MGEA 438 EUR
APOSOR12476-500MG
3-Phenyl-5,6-dihydroimidazo[2,1-b][1,3]thiazole-2-carbaldehyde
3-Phenyl-5,6-dihydroimidazo[2,1-b][1,3]thiazole-2-carbaldehyde
Formel: C₁₂H₁₀N₂OS
Molecular Weight: 230,29 g/mol
Schmelzpunkt: 138…139 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09817450
CAS-Nummer: 952182-40-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR