1-(3,4-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde

Lieferant: Apollo Scientific

OR12487-1G
APOSOR12487-1GEA 538 EUR
APOSOR12487-1G
1-(3,4-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
1-(3,4-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Formel: C₁₃H₉Cl₂NO₂
Molecular Weight: 282,13 g/mol
Schmelzpunkt: 144…146 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09817435
CAS-Nummer: 952183-56-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR