2-(4-Methoxyphenyl)pyrrolidine 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(2-Pyrrolidinyl)anisole
OR12504-1G
APOSOR12504-1GEA
128
EUR
APOSOR12504-1G
2-(4-Methoxyphenyl)pyrrolidine 95%
2-(4-Methoxyphenyl)pyrrolidine
Formel:
C₁₁H₁₅NO Molecular Weight: 177,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631836 CAS-Nummer: 74190-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...