2-(3,5-Dichlorophenyl)pyrrolidine

Lieferant: Apollo Scientific
Achtung

OR12506-250MG
APOSOR12506-250MGEA 0 EUR
APOSOR12506-250MG
2-(3,5-Dichlorophenyl)pyrrolidine
2-(3,5-Dichlorophenyl)pyrrolidine
Formel: C₁₀H₁₁Cl₂N
Lagertemperatur: Kühlschrank
MDL: MFCD02684093
CAS-Nummer: 887344-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR