L-Azetidin-2-Carboxylsäure

Lieferant: Apollo Scientific
Achtung

OR1250T-1G
APOSOR1250T-1GEA 0 EUR
APOSOR1250T-1G
L-Azetidin-2-Carboxylsäure
L-Azetidine-2-carboxylic acid

A four-membered ring analog of L-Proline used as an intermediate in the synthesis of polypeptides.

Formel: C₄H₇NO₂
Schmelzpunkt: 206…207 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005166
CAS-Nummer: 2133-34-8
EINECS: 218-362-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR