4-(2-Pyrrolidinyl)pyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Pyrrolidin-2-ylpyridine

OR12517-250MG
APOSOR12517-250MGEA 33.5 EUR
APOSOR12517-250MG
4-(2-Pyrrolidinyl)pyridin
4-(2-Pyrrolidinyl)pyridin
Formel: C₉H₁₂N₂
Molecular Weight: 148,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01862536
CAS-Nummer: 128562-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR