3-(Chloromethyl)-5-(3,5-dimethylisoxazol-4-yl)-1,2,4-oxadiazole

Lieferant: Apollo Scientific
Achtung

OR1253-250MG
APOSOR1253-250MGEA 37.7 EUR
APOSOR1253-250MG
3-(Chloromethyl)-5-(3,5-dimethylisoxazol-4-yl)-1,2,4-oxadiazole
3-(Chloromethyl)-5-(3,5-dimethylisoxazol-4-yl)-1,2,4-oxadiazole
Formel: C₈H₈ClN₃O₂
Molecular Weight: 213,621 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052098
CAS-Nummer: 175205-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR