2,6-Dichloroquinoline-3-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

OR12545-1G OR12545-250MG OR12545-5G
APOSOR12545-1GEA 317 EUR
APOSOR12545-1G APOSOR12545-250MG APOSOR12545-5G
2,6-Dichloroquinoline-3-carboxaldehyde
2,6-Dichloroquinoline-3-carboxaldehyde
Formel: C₁₀H₅Cl₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD06202347
CAS-Nummer: 73568-41-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR