trans-3-Amino-4-phenylpyrrolidine, N1-BOC protected

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl trans-3-amino-4-phenylpyrrolidine-1-carboxylate, trans-3-Amino-1-(tert-butoxycarbonyl)-4-phenylpyrrolidine

OR12547-100MG
APOSOR12547-100MGEA 77 EUR
APOSOR12547-100MG
trans-3-Amino-4-phenylpyrrolidine, N1-BOC protected
trans-3-Amino-4-phenylpyrrolidine, N1-BOC protected
Formel: C₁₅H₂₂N₂O₂
Molecular Weight: 262.347 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03002045
CAS-Nummer: 1015070-53-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR