3-Benzylpyrrolidine

Lieferant: Apollo Scientific
Achtung

OR12550-1G OR12550-250MG
APOSOR12550-1GEA 625 EUR
APOSOR12550-1G APOSOR12550-250MG
3-Benzylpyrrolidine
3-Benzylpyrrolidine
Formel: C₁₁H₁₅N
Lagertemperatur: Kühlschrank
MDL: MFCD01862537
CAS-Nummer: 170304-83-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR